Learn More
YMC America, Inc Cell-C col 3um 75x3mm
SDP
Catalog No. 5000878654
Encompass_Preferred
YMC s CHIRAL ART series features polysaccharide-derived chiral stationary phases (CSPs) based on amylose and cellulose tris-carbamate derivatives available in both coated and immobilized formats These columns provide exceptional stereoselectivity durability and solvent versatility supporting normal-phase reversed-phase and supercritical fluid chromatography (SFC) modes
Catalog No. 50-008-78654 Supplier YMC America, Inc Supplier No. KCN99S03L503WT
Add to Cart
Add to Cart
YMC s CHIRAL ART series features polysaccharide-derived chiral stationary phases (CSPs) based on amylose and cellulose tris-carbamate derivatives available in both coated and immobilized formats These columns provide exceptional stereoselectivity durability and solvent versatility supporting normal-phase reversed-phase and supercritical fluid chromatography (SFC) modes