Learn More
Hach Company Cell Cap for 5500sc Silica and Phosphate Analyzers (Replacement spare for colorimeter cell)
Catalog No. NC3360059 Shop All Hach Company Products
Encompass
Hach cell cap (MPN 6767800) is a replacement cap designed for the 5500sc silica and phosphate analyzers. It covers and helps protect the analyzer's colorimeter cell and is intended as a maintenance spare for replacement when the original cap is damaged or worn.
  • Replacement cap for colorimeter cell.
  • Helps protect optical surfaces from contamination.
  • Intended as a maintenance spare for affected analyzers.
Catalog No. NC3360059 Supplier Hach Company Supplier No. 6767800
Hach cell cap (MPN 6767800) is a replacement cap designed for the 5500sc silica and phosphate analyzers. It covers and helps protect the analyzer's colorimeter cell and is intended as a maintenance spare for replacement when the original cap is damaged or worn.
  • Replacement cap for colorimeter cell.
  • Helps protect optical surfaces from contamination.
  • Intended as a maintenance spare for affected analyzers.