Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cell Capture Imaging Reagent

Catalog No. LMR001MI Shop All MilliporeSigma Products
Change view
Click to view available options
Capacity:
8 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
LMR001MI 8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LMR001MI Supplier MilliporeSigma™ Supplier No. LMR001
Add to Cart
Add to Cart

Specially formulated polymeric hydrogel allowing cells to be constrained at the bottom of microtiter plates for subsequent imaging.

  • Specially formulated photopolymeric hydrogel thin film is optically transparent and permeable to antibodies and other immunocytochemistry reagents
  • Allows cells to be physically constrained at the bottom of standard imaging microtiter without any cell loss
  • Suitable for use in experiments where the cell population is less than 10,000
  • Cells captured can be easily micrographed regardless of the staining reagents
  • Displays sufficient mechanical strength to withstand routine pipetting and is dense enough for cells to sink to imaging surface
  • Uses:
    • Immunocytochemistry analysis
    • Use of this reagent with an antibody detected Jurkat cells
  • TRUSTED_SUSTAINABILITY

    Specifications

    Capacity 8 mL
    Content And Storage Stable for 1 yr (at 2°C to 8°C from date of receipt)
    Product Type Imaging Reagent
    For Use With (Application) Allows Cells to be Constrained at Bottom of Microtiter Plates for Subsequent Imaging, Immunocytochemistry Anaysis
    Form Liquid

    For research purposes only

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.