Learn More
Perkin Elmer US LLC Cell Changer for High Performance LAMBDAs
Catalog No. 502112892
Encompass_Preferred
The HP LAMBDA Linear Cell Changer is compatible with the LAMBDA 650/750/850/850+/950/1050/1050+ spectrometers.
Catalog No. 50-211-2892 Supplier Perkin Elmer US LLC Supplier No. L6020360
Add to Cart
Add to Cart
Request a Quote
The HP LAMBDA Linear Cell Changer is compatible with the LAMBDA 650/750/850/850+/950/1050/1050+ spectrometers.