Learn More
Cole-Parmer Cole-Parmer Conductivity Cell, K=1.0, Epoxy, Dip-Style, 100 Ohm Pt RTD ATC
Catalog No. 501961246 Shop All Cole-Parmer Products
Encompass_Preferred
Cells withstand pressures up to 150 psi (at 135C for SS; 25C for CPVC and PVDF).
Catalog No. 50-196-1246 Supplier Cole-Parmer Supplier No. 3582062
Cells withstand pressures up to 150 psi (at 135C for SS; 25C for CPVC and PVDF).