Learn More
Dojindo Molecular Technologies Inc Cell Count Normalization Kit (1000 TESTS) are used in cell number determination. No need for lysis procedure, can be used via plate reader.
SDP
Catalog No. NC1608117
Encompass
Cell Count Normalization Kit employs a nucleic acid staining dye, Hoechst 33342 which binds to nuclear DNA to emit blue fluorescence. Measuring the blue fluorescence allows for correction of the measured value that can easily be carried out in simple steps whereas the visual cell counting method requires cumbersome procedure. Moreover, unlike the correction by protein or ATP amount, the kit requires no lysis procedure. In addition, Quenching Buffer included in the kit enables a direct measuring of fluorescence signal without any background.
Catalog No. NC1608117 Supplier Dojindo Molecular Technologies Inc Supplier No. C54410
Cell Count Normalization Kit employs a nucleic acid staining dye, Hoechst 33342 which binds to nuclear DNA to emit blue fluorescence. Measuring the blue fluorescence allows for correction of the measured value that can easily be carried out in simple steps whereas the visual cell counting method requires cumbersome procedure. Moreover, unlike the correction by protein or ATP amount, the kit requires no lysis procedure. In addition, Quenching Buffer included in the kit enables a direct measuring of fluorescence signal without any background.