Learn More
Medicus Health Cell Counting Devices, 8 Key Differntial Counter
SDP
Catalog No. NC2012094
Encompass
∙ 8 key differential counter designed for manual white cell differential with white cell maturity pictograms ∙Eight cell counter great for manual differentials∙Signal bell sounds when 100 cells have been counted∙Wide base with rubber feet for stability∙Includes pitco-strip and totalizer window∙Features dual reset knobs to set counter back to zero∙Heavy duty impact resistant all plastic housing is chemical-resistant and easy to clean & disinfect∙Dimensions: 12.5"W x 3"D x 2"H
This item is not returnable. View return policy
Catalog No. NC2012094 Supplier Medicus Health Supplier No. 5267M6
∙ 8 key differential counter designed for manual white cell differential with white cell maturity pictograms ∙Eight cell counter great for manual differentials∙Signal bell sounds when 100 cells have been counted∙Wide base with rubber feet for stability∙Includes pitco-strip and totalizer window∙Features dual reset knobs to set counter back to zero∙Heavy duty impact resistant all plastic housing is chemical-resistant and easy to clean & disinfect∙Dimensions: 12.5"W x 3"D x 2"H