Learn More
Med Vet International Cell Counter, 8 key, J0505B
Catalog No. 5000584472
Encompass_Preferred
Differential Cell Counter allows the percentage of each type of white blood cells to be counted (leukocytes). Every stroke makes a distinct sound, and when a total of 100 is reached a bell will ring.
Catalog No. 50-005-84472 Supplier Med Vet International Supplier No. JOR505B
Differential Cell Counter allows the percentage of each type of white blood cells to be counted (leukocytes). Every stroke makes a distinct sound, and when a total of 100 is reached a bell will ring.