Learn More
Bio Rad Laboratories CELL COUNTING KIT 30 SLIDES
Catalog No. NC1723879
Encompass
30 slide pack of dual-chamber slides (60 counts), includes 1.5 ml trypan blue, for use with TC10™ or TC20™ automated cell counter
Catalog No. NC1723879 Supplier Bio Rad Laboratories Supplier No. 1450003
30 slide pack of dual-chamber slides (60 counts), includes 1.5 ml trypan blue, for use with TC10™ or TC20™ automated cell counter