Learn More
Enzo Life Sciences Cell Counting Kit-8 (500tests)
SDP
Catalog No. 502010100
Encompass_Preferred
The Cell Counting Kit-8 is a colorimetric assay kit used to measure cell proliferation and cytotoxicity. It is a ready-to-use solution that does not require radioisotopes and correlates with the [3H]-thymidine incorporation assay. It can be added directly to the cell media for fast, high-throughput screening without a solubilization process obtaining highly reproducible and accurate results. CCK-8 has shown to achieve higher sensitivity and stability than MTT, MTS or WST-1. Alternative name: CCKi-8. Applications: Colorimetric detection .
Catalog No. 50-201-0100 Supplier Enzo Life Sciences Supplier No. ALX850039KI02
The Cell Counting Kit-8 is a colorimetric assay kit used to measure cell proliferation and cytotoxicity. It is a ready-to-use solution that does not require radioisotopes and correlates with the [3H]-thymidine incorporation assay. It can be added directly to the cell media for fast, high-throughput screening without a solubilization process obtaining highly reproducible and accurate results. CCK-8 has shown to achieve higher sensitivity and stability than MTT, MTS or WST-1. Alternative name: CCKi-8. Applications: Colorimetric detection .