Learn More
Abcam Cell Counting Kit 8 (WST-8 / CCK8)
Supplier: Abcam AB228554
Cell Counting Kit 8 / CCK-8 Assay Kit / WST-8 Assay Kit ab228554 is a no-wash, mix-and-read, cell viability assay. The ready-to-use solution is added to cell cultures for a 1-4 hour incubation, with a colorimetric (460 nm) readout proportional to the number of living cells.- CCK-8 / WST-8 tetrazolium is reduced by cellular dehydrogenases in living cells and has very low cytotoxicity- The CCK-8 assay has higher sensitivity than other tetrazolium salt-based assays, such as MTT, MTS or XTT- Highly cited in >200 publications