Learn More
Enzo Life Sciences Cell Counting Kit-F (500 tests)
SDP
Catalog No. 502010132
Encompass_Preferred
Fluorometric kit for simple and accurate cell proliferation and cytotoxicity assays. Advantages: No radioisotopes, more sensitive than the colorimetric assay, short staining reaction time, no solubilization step, correlates with the [3H]-thymidine incorporation assay. Alternative name: CCKi-F. Applications: Fluorescence microscopy .
Catalog No. 50-201-0132 Supplier Enzo Life Sciences Supplier No. ALX850245KI02
Fluorometric kit for simple and accurate cell proliferation and cytotoxicity assays. Advantages: No radioisotopes, more sensitive than the colorimetric assay, short staining reaction time, no solubilization step, correlates with the [3H]-thymidine incorporation assay. Alternative name: CCKi-F. Applications: Fluorescence microscopy .