Learn More
Cole-Parmer Celline CL1000 Cell Culture System, One 1000 mL Media Reservoir
Catalog No. NC9707904 Shop All Cole-Parmer Products
Encompass
The cell culture systems feature separate growth and media chambers which provide continuous supply of nutrients and direct oxygenation. The systems also eliminate waste buildup for high cell densities.
This item is not returnable. View return policy
Catalog No. NC9707904 Supplier Cole-Parmer Supplier No. 1305220
The cell culture systems feature separate growth and media chambers which provide continuous supply of nutrients and direct oxygenation. The systems also eliminate waste buildup for high cell densities.