Learn More
BIOLOGIX USA INC CELL CULTR DISH 60X20MM W/STIR
Catalog No. NC2445684
Encompass_Preferred
Weight: 0.88 lbs▪ Mesh membrane material: Nylon▪Frame material: PS▪ Mesh membrane thickness: 10 ~ 23 μm▪ Membrane pore size: 0.40 μm
Catalog No. NC2445684 Supplier BIOLOGIX USA INC Supplier No. 911607
Weight: 0.88 lbs▪ Mesh membrane material: Nylon▪Frame material: PS▪ Mesh membrane thickness: 10 ~ 23 μm▪ Membrane pore size: 0.40 μm