Learn More
Alkali Scientific Cell Culture Chamber Slides, 8 Wells, PS/PP, Glass Slide, Sterile, 12/cs
SDP
Catalog No. 502336170
Encompass_Preferred
ASI chamber slides are designed specifically to cover all your cell-based microscopic research needs and use a patented design that simplifies your work. Our state-of-the-art slides allow you to grow cell cultures within each chamber, making it possible for you to prepare a higher number of samples on a single glass slide. The volume of each well is 1.2 mL for the 8 wells culture slides. The chambers are produced from high quality clear plastic, and each of the chambers is marked with an alphanumerical code which makes tracking the sample easier. They are sterilized through the use of gamma irradiation, ensuring that this product is ready for use as soon as you receive it. They are also RNase, DNase and Pyrogen free. Materials: Chamber PS / Slide Glass / Holder PP.
Catalog No. 50-233-6170 Supplier Alkali Scientific Supplier No. TCC008
ASI chamber slides are designed specifically to cover all your cell-based microscopic research needs and use a patented design that simplifies your work. Our state-of-the-art slides allow you to grow cell cultures within each chamber, making it possible for you to prepare a higher number of samples on a single glass slide. The volume of each well is 1.2 mL for the 8 wells culture slides. The chambers are produced from high quality clear plastic, and each of the chambers is marked with an alphanumerical code which makes tracking the sample easier. They are sterilized through the use of gamma irradiation, ensuring that this product is ready for use as soon as you receive it. They are also RNase, DNase and Pyrogen free. Materials: Chamber PS / Slide Glass / Holder PP.