Learn More
Alkali Scientific ASI™ Cell Culture Dish, 35 X 10mm, 3mL*, TC Treated, Sterile, 10/sleeve, 500/cs
SDP
Catalog No. 502336082
Encompass_Preferred
ASI™ Cell Culture Dishes are optically clear for microscopy with large pronounced gripping rings and air exchange vents. They are pyrogen free, Gamma sterilized, and USP guideline compliant (USP class VI tested). Tissue culture treated.
Catalog No. 50-233-6082 Supplier Alkali Scientific Supplier No. TD0035
ASI™ Cell Culture Dishes are optically clear for microscopy with large pronounced gripping rings and air exchange vents. They are pyrogen free, Gamma sterilized, and USP guideline compliant (USP class VI tested). Tissue culture treated.