Learn More
Alkali Scientific ASI™ Cell Culture Dish, 60 X 15mm, 5mL *, TC Treated, Sterile, 10/Sleeve, 500/cs
SDP
Catalog No. 502335775
Encompass_Preferred
ASI™ Cell Culture Dishes are optically clear for microscopy with large pronounced gripping rings and air exchange vents. They are pyrogen free, Gamma sterilized, and USP guideline compliant (USP class VI tested). Tissue culture treated.
Catalog No. 50-233-5775 Supplier Alkali Scientific Supplier No. TD0060
ASI™ Cell Culture Dishes are optically clear for microscopy with large pronounced gripping rings and air exchange vents. They are pyrogen free, Gamma sterilized, and USP guideline compliant (USP class VI tested). Tissue culture treated.