Learn More
Alkali Scientific Cell Culture Plate, PS, 96-Well, Black Wells with Clear Bottoms, Flat Bottom, TC Treated, with Lid, 5/Sterile Inner Pack, 25/cs
SDP
Catalog No. 502335911
Encompass_Preferred
Ideal plates for obtaining amplified signals in fluorescence assay while minimizing cross-talk utilizing the light blocking benefit of a black plate. The highly transparent bottom provides an optimal environment for microscopic observation. Ultra clear bottom for Cell Imaging on all instrumentation Optimized surface technology for cell attachment and microscopic observation. Tissue Culture Treated for cell adhesion and attachment providing an ideal proliferation environment. ▪ Black Wall and topside (PS) / Clear Bottom ▪ Bottom material: Optically Clear PS ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Alphanumeric labeling ▪ Provided with matching low evaporation lid ▪ sterile 5/pack ▪ Non – pyrogenic ▪ Non – cytotoxic ▪ DNase / RNase – free ▪ Human DNA – free
Catalog No. 50-233-5911 Supplier Alkali Scientific Supplier No. TPB1096
Add to Cart
Add to Cart
Ideal plates for obtaining amplified signals in fluorescence assay while minimizing cross-talk utilizing the light blocking benefit of a black plate. The highly transparent bottom provides an optimal environment for microscopic observation. Ultra clear bottom for Cell Imaging on all instrumentation Optimized surface technology for cell attachment and microscopic observation. Tissue Culture Treated for cell adhesion and attachment providing an ideal proliferation environment. ▪ Black Wall and topside (PS) / Clear Bottom ▪ Bottom material: Optically Clear PS ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Alphanumeric labeling ▪ Provided with matching low evaporation lid ▪ sterile 5/pack ▪ Non – pyrogenic ▪ Non – cytotoxic ▪ DNase / RNase – free ▪ Human DNA – free