Learn More
Alkali Scientific Cell Culture Plate, PS, 96-Well, Black, Flat Bottom, TC Treated, with Lid, Individually Wrapped, 50/cs
SDP
Catalog No. 502336081
Encompass_Preferred
ASI™ 96 Well Culture Solid Black Plates have a low auto-fluorescence with efficient light blocking ability to obtain most accurate result in fluorescence experiments. Black microplates have low background fluorescence and minimize light scattering. ▪ High Throughput Screening compatible ▪ Suitable for fluorescence & luminescence assays ▪ Non treated for suspension cultures ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Molded Alphanumeric labeling ▪ USP guideline compliant (USP class Ⅵ tested) ▪ ANSI guideline compliant ▪ Non pyrogenic ▪ Non cytotoxic ▪ DNase / RNase / Pyrogen free ▪ Human DNA free
Catalog No. 50-233-6081 Supplier Alkali Scientific Supplier No. TPB9096
Add to Cart
Add to Cart
ASI™ 96 Well Culture Solid Black Plates have a low auto-fluorescence with efficient light blocking ability to obtain most accurate result in fluorescence experiments. Black microplates have low background fluorescence and minimize light scattering. ▪ High Throughput Screening compatible ▪ Suitable for fluorescence & luminescence assays ▪ Non treated for suspension cultures ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Molded Alphanumeric labeling ▪ USP guideline compliant (USP class Ⅵ tested) ▪ ANSI guideline compliant ▪ Non pyrogenic ▪ Non cytotoxic ▪ DNase / RNase / Pyrogen free ▪ Human DNA free