Learn More
Chemglass Life Sciences Bag, Cell Culture, Motion, 10L, Ultra Clear, CellGrow
SDP
Catalog No. NC1563981
Encompass
Cell culture bags are manufactured from ultra clear Flex 200 robust film and gamma irradiated.The film meets USP Class VI specifications and is contamination free.The inner product contact material is low density polyethylene.All bags and components content materials are USP Class VI tested.Excellent choice for mammalian and plant culture cell lines with extended mixing times.   
Catalog No. NC1563981 Supplier Chemglass Life Sciences Supplier No. CLS120710M
Cell culture bags are manufactured from ultra clear Flex 200 robust film and gamma irradiated.The film meets USP Class VI specifications and is contamination free.The inner product contact material is low density polyethylene.All bags and components content materials are USP Class VI tested.Excellent choice for mammalian and plant culture cell lines with extended mixing times.   