Learn More
Alkali Scientific ASI™ Cell Culture Chamber Slide, Clear, D-Lux +Surface, PS & PP, Sterile, 12/Case, 4 Well
SDP
Catalog No. 503028774
Encompass_Preferred
Cell Culture Chamber Slides are designed for various cell-based experiments and are also useful for virus, toxicity tests and immunocytological experiments. Cell Culture Chamber Slides provide removable chamber that enables cell growth & imaging directly on the slide itself, convenient for staining and microscopic examination without cell transfer.  This process makes Alkali's chamber slides perfect for delicate or unstable cell types. Chambers with various well numbers and colors are available for different applications. With new additions of novel culture surfaces DLux & FLux we at Alkali Scientific are pleased to expand your surface type options.   Surface materials for Cell Culture SlideGlass: naturally charged, good growth surface for adherent cellsDLux: surface modified plastic with minimal autofluorescenceFLux: surface modified plastic film (188 μm thick), optimal for confocal microscopy
Catalog No. 50-302-8774 Supplier Alkali Scientific Supplier No. TCD04
Cell Culture Chamber Slides are designed for various cell-based experiments and are also useful for virus, toxicity tests and immunocytological experiments. Cell Culture Chamber Slides provide removable chamber that enables cell growth & imaging directly on the slide itself, convenient for staining and microscopic examination without cell transfer.  This process makes Alkali's chamber slides perfect for delicate or unstable cell types. Chambers with various well numbers and colors are available for different applications. With new additions of novel culture surfaces DLux & FLux we at Alkali Scientific are pleased to expand your surface type options.   Surface materials for Cell Culture SlideGlass: naturally charged, good growth surface for adherent cellsDLux: surface modified plastic with minimal autofluorescenceFLux: surface modified plastic film (188 μm thick), optimal for confocal microscopy