Learn More
Alkali Scientific Cell Culture Chamber Slides 12
SDP
Catalog No. 503028854
Encompass_Preferred
ASI cell culture chamber slides are designed specifically to cover all your cell-based microscopic research needs and use a patented design that simplifies your work Our state-of-the-art slides allow you to grow cell cultures within each chamber making it possible for you to prepare a higher number of samples on a single glass slide Choose from1 2 4 8 or 12 wells The volume of each well is 1 2 mL for the 8 wells culture slides and 2 5mL for the 4 wells cell culture slides The chambers are produced from high quality clear plastic and each of the chambers is marked with an alphanumerical code which makes tracking the sample easier They are sterilized through the use of gamma irradiation ensuring that this product is ready for use as soon as you receive it They are also RNase DNase and Pyrogen free Materials Chamber PS / Slide Glass / Holder PP 1 2 4 8 and 12 Well Slides Available Alphanumeric Well Identification Silicone gasket provides leak-free operation Oversized hinged clasps
Catalog No. 50-302-8854 Supplier Alkali Scientific Supplier No. TCC0012
ASI cell culture chamber slides are designed specifically to cover all your cell-based microscopic research needs and use a patented design that simplifies your work Our state-of-the-art slides allow you to grow cell cultures within each chamber making it possible for you to prepare a higher number of samples on a single glass slide Choose from1 2 4 8 or 12 wells The volume of each well is 1 2 mL for the 8 wells culture slides and 2 5mL for the 4 wells cell culture slides The chambers are produced from high quality clear plastic and each of the chambers is marked with an alphanumerical code which makes tracking the sample easier They are sterilized through the use of gamma irradiation ensuring that this product is ready for use as soon as you receive it They are also RNase DNase and Pyrogen free Materials Chamber PS / Slide Glass / Holder PP 1 2 4 8 and 12 Well Slides Available Alphanumeric Well Identification Silicone gasket provides leak-free operation Oversized hinged clasps