Learn More
Electron Microscopy Sciences Costar Cell Culture Clusters 24 well
SDP
Supplier:  Electron Microscopy Sciences 6470705
Encompass_Preferred
Tissue culture plates are designed and treated for a wide range of applications. Plates are made from optically clear, medical grade polystyrene with flat bottoms of uniform thickness, allowing for distortion-free observation and microscopic examination. Alphanumeric coordinates are placed on the same focal plane as cell growth-for easy referencing of well position. These plates are also for use with our NetWells and our MeshWell Plates.
Catalog No. 50-310-68