Learn More
Electron Microscopy Sciences Secureslip™ Cell Culture Coverglass 15 well, 12 mm dia
SDP
Catalog No. 5031007
Encompass_Preferred
Convenient and secure coverglass that will not move during cultivation. The coverglass is affixed to a thin microscopically transparent silicone base which secures to culture vessels by an electrostatic charge, not an adhesive, to prevent movement during cultivation.
The silicone also acts as a hydrophobic barrier to isolate liquid reagents on glass. Individual coverglass with silicone backing can be adhered to a petri dish or plate. These can be easily removed one at a time from the dish during culture. Once removed, coverglass can be affixed to a microscope slide by means of the tacky silicone backing for routine processing, or placed cell side down onto a drop of reagent using the silicone backing as a cover to prevent evaporation.
Size: 12 mm diameter
Catalog No. 50-310-07 Supplier Electron Microscopy Sciences Supplier No. 7046214
Convenient and secure coverglass that will not move during cultivation. The coverglass is affixed to a thin microscopically transparent silicone base which secures to culture vessels by an electrostatic charge, not an adhesive, to prevent movement during cultivation.
The silicone also acts as a hydrophobic barrier to isolate liquid reagents on glass. Individual coverglass with silicone backing can be adhered to a petri dish or plate. These can be easily removed one at a time from the dish during culture. Once removed, coverglass can be affixed to a microscope slide by means of the tacky silicone backing for routine processing, or placed cell side down onto a drop of reagent using the silicone backing as a cover to prevent evaporation.
Size: 12 mm diameter