Learn More
Electron Microscopy Sciences Multislip™ Cell Culture Coverglass Insert, 8-18 x 18 mm
SDP
Catalog No. 50930319
Encompass_Preferred
Sterile, ready-to-use MultiSlip for culture of cells where pretreatment with a biological coating of glass surface is required. MultiSlip insert number 1.5 thickness German coverglass per insert are sterile and ready to use in conventional 86 mm x 128 mm culture plates.
Staining and washing procedures may be performed with MultiSlip inserts in the plate, or silicone backed coverglass may be removed individually and affixed to glass microscope slides. Alternatively, inserts may be easily removed for batch processing in glass staining dishes.
MultiSlips are ideally suited for the culture of cells where pretreatment of glass surfaces with a biological coating is required. Simply add sterile solution to the plate incubates and aspirate. Coating is applied evenly to one side of the glass only, with no over-lapping, handling with forceps.
Size: 8-18 x 18 mm
Catalog No. 50-930-319 Supplier Electron Microscopy Sciences Supplier No. 7046318
Sterile, ready-to-use MultiSlip for culture of cells where pretreatment with a biological coating of glass surface is required. MultiSlip insert number 1.5 thickness German coverglass per insert are sterile and ready to use in conventional 86 mm x 128 mm culture plates.
Staining and washing procedures may be performed with MultiSlip inserts in the plate, or silicone backed coverglass may be removed individually and affixed to glass microscope slides. Alternatively, inserts may be easily removed for batch processing in glass staining dishes.
MultiSlips are ideally suited for the culture of cells where pretreatment of glass surfaces with a biological coating is required. Simply add sterile solution to the plate incubates and aspirate. Coating is applied evenly to one side of the glass only, with no over-lapping, handling with forceps.
Size: 8-18 x 18 mm