Learn More
Mascon Medical Mascon Medical™ Cell Culture Dish, 100mm, 10EA/Bag, 30 Bags/Case
SDP
Catalog No. NC2458293
Encompass_Preferred
Mascon Medical™ Cell Culture Dishes are the best choice for direct and convenient cell processing. The cell culture dish has a wave-like easy-to-grip ring to ensure safety when taking the culture dish or cell operation. Features:•TC treatment•Sterilization assurance level: SAL 10-6•Meets ISO13485 quality management system standardSpecifications:•Volume: 100mm•Qty. / Case: 300•Color: Transparent•Material: Polystyrene•Size: Φ94*21.5mm•Packaging Format: 10EA/Bag, 30Bags/Case
Catalog No. NC2458293 Supplier Mascon Medical Supplier No. FSCCDISH
Mascon Medical™ Cell Culture Dishes are the best choice for direct and convenient cell processing. The cell culture dish has a wave-like easy-to-grip ring to ensure safety when taking the culture dish or cell operation. Features:•TC treatment•Sterilization assurance level: SAL 10-6•Meets ISO13485 quality management system standardSpecifications:•Volume: 100mm•Qty. / Case: 300•Color: Transparent•Material: Polystyrene•Size: Φ94*21.5mm•Packaging Format: 10EA/Bag, 30Bags/Case