Learn More
Chemglass Life Sciences Cell Culture Dish, 35mm, Gripping Ring, TC, Sterile, 20/Pk, 500/Case
SDP
Catalog No. 501897807
Encompass_Preferred
NEST cell culture dishes are ideal for all types of culturing where microscopic observation is required. Numeric marks on the bottom of each dish allow users to identify the location of cells.High clarity polystyreneFlat transparent surface for distortion-free observationVacuum plasma TC treatment, excellent cell adherenceStackable for easy storage and handlingSterilized by E-BeamNon-pyrogenic
Catalog No. 50-189-7807 Supplier Chemglass Life Sciences Supplier No. CGN1805035GR
NEST cell culture dishes are ideal for all types of culturing where microscopic observation is required. Numeric marks on the bottom of each dish allow users to identify the location of cells.High clarity polystyreneFlat transparent surface for distortion-free observationVacuum plasma TC treatment, excellent cell adherenceStackable for easy storage and handlingSterilized by E-BeamNon-pyrogenic