Learn More
Stellar Scientific CELL CULTURE FLASK 200/CS
SDP
Catalog No. NC2308947
Encompass
T-25 25cm2 Polystyrene Cell Culture Flask Vent Cap Non-Treated Surface RNase and DNase Free STERILE 10/PK 200/CS
Catalog No. NC2308947 Supplier Stellar Scientific Supplier No. TC30210
T-25 25cm2 Polystyrene Cell Culture Flask Vent Cap Non-Treated Surface RNase and DNase Free STERILE 10/PK 200/CS