Learn More
Research Products International Corp Cell Culture Flask, 75 cm2, Sterile, with Plug Cap, 100 per Case
SDP
Catalog No. 501367609
Encompass
Manufactured from ultra clear polystyrene for easy viewing of contents. Proprietary surface treating ensures optimum cell attachment and growth. Additional features include graduated markings on both sides of flask, angled neck design. Supplied sterile, non-pyrogenic, RNase, DNase free and 5 flasks per bag. The flasks are packaged in clear plastic bags.
Plug cap flasks:
Suitable for cell and tissue culture under sealed conditions, completely isolating the culture from the outside environment.
Catalog No. 50-136-7609 Supplier Research Products International Corp Supplier No. 162466P
Manufactured from ultra clear polystyrene for easy viewing of contents. Proprietary surface treating ensures optimum cell attachment and growth. Additional features include graduated markings on both sides of flask, angled neck design. Supplied sterile, non-pyrogenic, RNase, DNase free and 5 flasks per bag. The flasks are packaged in clear plastic bags.
Plug cap flasks:
Suitable for cell and tissue culture under sealed conditions, completely isolating the culture from the outside environment.