Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cell Culture Flask, Vent Cap, TC-treated

Catalog No. MP096142030 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Culture Area:
175 cm²
25 cm²
75 cm²
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area
MP096142030 175 cm²
MP096140200 25 cm²
MP096141100 75 cm²
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP096142030 Supplier MP Biomedicals™ Supplier No. 096142030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell culture flasks are engineered for culture efficiency to assure that results of research are reproducible, as well as reliable in consistency.

  • Available with treated surface, suitable for adherent cell culture, suitable for suspension cell culture
  • Available in three sizes: 25, 75, and 175 cm² with a flat surface (bottom of flask)
  • Mouth design that gives easy access to entire surface of flask
  • Graduated on both sides
  • Includes white blank area for labeling
  • Flasks are designed to stack easily for best use of incubator space
  • Sterilized by irradiation
  • Pyrogen-free
  • DNAse/RNase Free
TRUSTED_SUSTAINABILITY

Specifications

Closure Color White
Material Polystyrene
For Use With (Application) Adherent cell culture, Cell expansion
Includes 30 x Flasks, 30 x Filtered Vented Caps
Product Type Cell Culture Flask
Writing Surface Yes
Graduated Yes
Sterility Sterile
Bottom Shape Flat
Closure Type Vent Cap
Culture Area 175 cm²
Closure Size 0.2 μm
Quantity 30/Cs.
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.