Learn More
Quality Biological Inc Water, Cell Culture, WFI Quality, 1000mL
SDP
Supplier:  Quality Biological Inc 118162131
Encompass_Preferred
QBI’s Water, Cell Culture, WFI Quality is high quality water for use as a solvent in the preparation of cell culture media and biochemical reagents. This water is extensively purified by reverse osmosis, subsequently deionized and passed through a filter membrane to remove particles and tested to meet the specifications found in the USP (United States Pharmacopeia) monograph for Water for Injection (WFI) for endotoxin, TOC, and conductivity. It is additionally tested for sterility, mycoplasma and cell culture performance. This water is tested against USP specifications, but is not intended for injection. It is only intended for research or further manufacturing use, not for diagnostic or therapeutic use.Water, Cell Culture, WFI Quality is • Endotoxin less than 0.03 EU/mL• Mycoplasma and Sterility Tested• Growth Performance TestedStorage Condition: 15°C to 30°CShipping Condition: ambient temperatureMax Shelf Life: 24 months from date of manufacture
Catalog No. 50-983-221