Learn More
Eurofins Discoverx AssayComplete Cell Culture Kit-110, (92-3110G)
Catalog No. 501999855
Encompass
AssayComplete Cell Culture Kit-110 is an optimized media kit intended for use in culturing and propagating Eurofins DiscoverX cell lines. This kit contains the base medium, relevant serum, and pen/strep required to produce a full cell culture medium for optimal cell culture results. This product is provided as a 3-component kit which will produce approximately 550 mL of finished media. Please refer to cell line specific datasheet to determine the recommended cell culture kit. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. 50-199-9855 Supplier Eurofins Discoverx Supplier No. 923110G
AssayComplete Cell Culture Kit-110 is an optimized media kit intended for use in culturing and propagating Eurofins DiscoverX cell lines. This kit contains the base medium, relevant serum, and pen/strep required to produce a full cell culture medium for optimal cell culture results. This product is provided as a 3-component kit which will produce approximately 550 mL of finished media. Please refer to cell line specific datasheet to determine the recommended cell culture kit. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.