Learn More
STEMCELL Technologies PneumaCult™ Alveolar Organoid Differentiation Medium, Size: 1 Kit
SDP
Catalog No. 502250330
Encompass
Cell culture medium for differentiation of ATII alveolar organoids into ATI organoids for downstream applications
This item is not returnable. View return policy
Catalog No. 50-225-0330 Supplier STEMCELL Technologies Supplier No. 1000861
Cell culture medium for differentiation of ATII alveolar organoids into ATI organoids for downstream applications