Learn More
STEMCELL Technologies IntestiCult™ Organoid Differentiation Medium (Human)
SDP
Catalog No. 502045485
Encompass
IntestiCult™ Organoid Differentiation Medium (Human) supports differentiation of intestinal organoids in 3D or in 2D as monolayers or air-liquid interface cultures. Starting cultures include intestinal organoids derived from human intestinal crypts, or passaged organoids that have been cultured with IntestiCult™ Organoid Growth Medium (Human, Catalog #06010).
Catalog No. 50-204-5485 Supplier STEMCELL Technologies Supplier No. 1000214
IntestiCult™ Organoid Differentiation Medium (Human) supports differentiation of intestinal organoids in 3D or in 2D as monolayers or air-liquid interface cultures. Starting cultures include intestinal organoids derived from human intestinal crypts, or passaged organoids that have been cultured with IntestiCult™ Organoid Growth Medium (Human, Catalog #06010).