Learn More
Beckman Coulter Qualified Reservoir, 2x3 Well, Polypropylene Microplate, Surface Treated with Amine Coating
Catalog No. NC1731998
Encompass
The Echo Qualified Reservoir is designed to enable convenient and affordable large volume transfers on the Echo 525 Liquid Handling System. Its 6-well design enables the transfer of over 15 mL from a single reservoir plate.
Catalog No. NC1731998 Supplier Beckman Coulter Supplier No. 00111101
The Echo Qualified Reservoir is designed to enable convenient and affordable large volume transfers on the Echo 525 Liquid Handling System. Its 6-well design enables the transfer of over 15 mL from a single reservoir plate.