Learn More
GenDEPOT Cell Culture Penicillin 100ml
Catalog No. 501959713
Encompass_Preferred
Contains 10,000 units of penicillin (base) and 10,000 µg of streptomycin (base)/ml utilizing penicillin G (sodium salt) and streptomycin sulfate in 0.85% saline.
Catalog No. 50-195-9713 Supplier GenDEPOT Supplier No. CA005010
Contains 10,000 units of penicillin (base) and 10,000 µg of streptomycin (base)/ml utilizing penicillin G (sodium salt) and streptomycin sulfate in 0.85% saline.