Learn More
Mascon Medical Mascon Medical™ Cell Culture Plate, 6-Well, Sterile, 1EA/Bag, 100 Bags/Case
SDP
Catalog No. NC2458300
Encompass_Preferred
Cell Culture Plate 6-WellMascon Medical™ 6-Well Cell Culture Plates are ideal for small-volume cell expansion cell-based screening. They are suitable for cell culture, cell transfection, immunofluorescence, clone formation, and other experiments. The cell culture plates ensure good optical detection performance and improve the convenience of manual or automated data and reading by improving the flatness of the culture surface, using highly transparent materials, and reducing the generation of a half-moon liquid surface in the culture hole. Features:•TC treatment•Flat bottom•Sterile (individually packed)•Ventilation holes for the exchange of gas and temperature•Anti-slip bottom•Alphanumeric border markings•Meets ISO13485 quality management system standardSpecifications:•Volume: 6-Well•Qty. / Case: 100•Color: Transparent•Material: Polystyrene•Size: 127.6*85.2*22.2mm•Packaging Format: 1EA/Bag, 100Bags/Case
Catalog No. NC2458300 Supplier Mascon Medical Supplier No. FSCCPLATE6
Cell Culture Plate 6-WellMascon Medical™ 6-Well Cell Culture Plates are ideal for small-volume cell expansion cell-based screening. They are suitable for cell culture, cell transfection, immunofluorescence, clone formation, and other experiments. The cell culture plates ensure good optical detection performance and improve the convenience of manual or automated data and reading by improving the flatness of the culture surface, using highly transparent materials, and reducing the generation of a half-moon liquid surface in the culture hole. Features:•TC treatment•Flat bottom•Sterile (individually packed)•Ventilation holes for the exchange of gas and temperature•Anti-slip bottom•Alphanumeric border markings•Meets ISO13485 quality management system standardSpecifications:•Volume: 6-Well•Qty. / Case: 100•Color: Transparent•Material: Polystyrene•Size: 127.6*85.2*22.2mm•Packaging Format: 1EA/Bag, 100Bags/Case