Learn More
Alkali Scientific Cell Culture Plate, flat, TC, sterile, Bulk, 12 Well, 10/pk, 50/cs
SDP
Catalog No. 503028915
Encompass_Preferred
Cell Culture Plates For Tissue Culture Treatment, Flat, Treated, Sterile High clarity, 100% virgin polystyrene •  Sterilized by E-beam•  Vacuum Plasma tissue culture treatment•  Clear lot number for batch traceability•  Peel-to-open, individually wrapped•  Markings of well coordinates available for 12, 24, 48, and 96 well plates•  Non-Pyrogenic, DNase/RNase Free  
Catalog No. 50-302-8915 Supplier Alkali Scientific Supplier No. TPN2012
Cell Culture Plates For Tissue Culture Treatment, Flat, Treated, Sterile High clarity, 100% virgin polystyrene •  Sterilized by E-beam•  Vacuum Plasma tissue culture treatment•  Clear lot number for batch traceability•  Peel-to-open, individually wrapped•  Markings of well coordinates available for 12, 24, 48, and 96 well plates•  Non-Pyrogenic, DNase/RNase Free  