Learn More
Alkali Scientific Cell Culture Plate PS 96 Well
SDP
Catalog No. 503028925
Encompass_Preferred
Whether culturing individual cell lines or scaling up for high-throughput screening our black plates with a clear bottom and sterile design are ideal for enhancing fluorescence assay signals while reducing cross-talk thanks to the light-blocking properties of the black plate Advancements in surface modifications well geometry and optical clarity have led to a range of 96-well cell culture plates tailored for specific applications including optical bottom plates designed for cellular imaging To ensure reliability flexibility and reproducibility at every stage of cell culture these plates are available with various surface treatments optimized for cell attachment and growth The highly transparent bottom offers an optimal environment for microscopic observation Tissue Culture Treated for cell adhesion and attachment providing an ideal proliferation environment Black Wall and topside (PS) / Clear Bottom Bottom material Optically Clear PS Designed to prevent cross contamination Eff
Catalog No. 50-302-8925 Supplier Alkali Scientific Supplier No. TPB2096
Whether culturing individual cell lines or scaling up for high-throughput screening our black plates with a clear bottom and sterile design are ideal for enhancing fluorescence assay signals while reducing cross-talk thanks to the light-blocking properties of the black plate Advancements in surface modifications well geometry and optical clarity have led to a range of 96-well cell culture plates tailored for specific applications including optical bottom plates designed for cellular imaging To ensure reliability flexibility and reproducibility at every stage of cell culture these plates are available with various surface treatments optimized for cell attachment and growth The highly transparent bottom offers an optimal environment for microscopic observation Tissue Culture Treated for cell adhesion and attachment providing an ideal proliferation environment Black Wall and topside (PS) / Clear Bottom Bottom material Optically Clear PS Designed to prevent cross contamination Eff