Learn More
Research Products International Corp Cell Culture Plates, 12 Well, 50 per Case
SDP
Catalog No. 501367496
Encompass
Cell Culture Plates are designed for a wide range of cell and tissue culture applications. Manufactured from crystal clear virgin polystyrene with proprietary surface treating to ensure optimum cell attachment and growth. Plates feature raised well rims, flat bottom, thin-wall well bottoms for thermal consistency and optimal viewing. Supplied with lids.
Catalog No. 50-136-7496 Supplier Research Products International Corp Supplier No. 141373
Add to Cart
Add to Cart
Cell Culture Plates are designed for a wide range of cell and tissue culture applications. Manufactured from crystal clear virgin polystyrene with proprietary surface treating to ensure optimum cell attachment and growth. Plates feature raised well rims, flat bottom, thin-wall well bottoms for thermal consistency and optimal viewing. Supplied with lids.