Learn More
Apexbio Technology LLC Cell Cycle Assay Kit(Synonyms: Cell Cycle Analysis Kit, PI Cell Cycle Kit, Cell Cycle Detection Kit, Propidium Iodide Cell Cycle Assay), 50T.
SDP
Catalog No. 5000061088
Encompass_Preferred
The Cell Cycle Assay Kit utilizes propidium iodide (PI) a nucleic acid-intercalating fluorescent dye to evaluate cell cycle progression based on cellular DNA content PI selectively stains fixed or membrane-compromised cells binding stoichiometrically to DNA Fluorescence intensity correlates directly with DNA amount distinguishing clearly between G0/G1 (2N) S (2N-4N) and G2/M (4N) phase cells Apoptotic cells exhibit reduced fluorescence (sub-G1 peaks) due to DNA fragmentation This kit enables precise analysis of cell cycle dynamics and apoptosis via flow cytometry supporting research in cell biology oncology and pharmacology
Catalog No. 50-000-61088 Supplier Apexbio Technology LLC Supplier No. K226350
Add to Cart
Add to Cart
The Cell Cycle Assay Kit utilizes propidium iodide (PI) a nucleic acid-intercalating fluorescent dye to evaluate cell cycle progression based on cellular DNA content PI selectively stains fixed or membrane-compromised cells binding stoichiometrically to DNA Fluorescence intensity correlates directly with DNA amount distinguishing clearly between G0/G1 (2N) S (2N-4N) and G2/M (4N) phase cells Apoptotic cells exhibit reduced fluorescence (sub-G1 peaks) due to DNA fragmentation This kit enables precise analysis of cell cycle dynamics and apoptosis via flow cytometry supporting research in cell biology oncology and pharmacology