Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Cell Cycle and Mitosis 3D Model Making Kit
SCI_ED

Catalog No. S436049 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S436049 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S436049 Supplier NewPath Learning™ Supplier No. 247742
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Cell Cycle and Mitosis 3D Model Making Kit allows students to make models of cells and chromosomes to learn about mitosis, the cell cycle, and the growth of subcellular structures and replication of DNA during interphase.

  • Introduce your students to the cell cycle and how it is divided into different stages, including mitosis.
  • Your students will begin by making models of cells and chromosomes, they then learn about what happens during mitosis and the cell cycle.
  • In the process, they will look at growth of subcellular structures and replication of DNA during interphase.
  • Next, they learn the steps of mitosis including cell nucleus break down, line up of chromosomes on cell fibers, chromosome separation and nucleus reformation and cytokinesis.
  • The Model Making Kit includes enough materials for students to assemble up to 5 paper models working cooperatively.
  • The comprehensive Teacher Resource Guide included provides illustrated readings, activities, key vocabulary and an assessment, along with teaching opportunities for students to Engage, Explore, Explain, Extend and Evaluate their understanding of the concepts presented.

Specifications

For Use With (Application) Model Assembly
Grade 6 to 10
Includes 5 Models, Teacher Resource Guide, Vinyl Pouch
Length (English) 12 in.
Material Paper
Weight (English) 2 lb.
Width (English) 9 in.
Product Type 3D Model
Grade Level Intermediate - Middle School, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.