Learn More
Neuromics, Inc Cell Detachment Solution
Catalog No. NC1553918
Encompass
Cell Detachment Solution is an effective alternative to Trypsin/EDTA for in vitro cultured cells, containing proteolytic and collagenolytic enzymes. Maximum cells viability. No need to wash detached cells. Gentle and rapid detachment. Effective on a wide range of cells.
Catalog No. NC1553918 Supplier Neuromics, Inc Supplier No. ADF001
Cell Detachment Solution is an effective alternative to Trypsin/EDTA for in vitro cultured cells, containing proteolytic and collagenolytic enzymes. Maximum cells viability. No need to wash detached cells. Gentle and rapid detachment. Effective on a wide range of cells.