Learn More
Promocell GmbH CELL DISSOC. SOL ACF 100ML
Supplier:  Promocell GmbH C41320
Encompass
Biological Buffers, PromoCell, Cell Dissociation Solution ACF, 100ML. Cell Dissociation Solution ACF is a proprietary non-enzymatic reagent for detachment of adherent cell types from tissue culture plastic or glass surfaces. Due to its gentle yet effective action it presents a good animal component-free alternative to trypsin and other enzymatic detachment solutions.
Catalog No. 50-306-393