Learn More
Sigma Aldrich Fine Chemicals Biosciences Cell Dissociation Solution
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C5789100ML
Encompass
Cell Dissociation SOLUTIONon Non-enzymatic buffer systems are alternative to the canonical trypsin based buffers for dissociation of adherent mesenchymal stem cell (MSC) monolayers.
Catalog No. 50-290-1636