Learn More
Research Products International Corp Cell Divider for Tube Storage Boxes, Cardboard, 12 x 12 Array, 9.6mm Opening, 144 Tube Capacity
SDP
Catalog No. 50998924
Encompass
Cardboard cell divider is designed for use with cardboard storage boxes and cardboard freezer boxes.
Cell divider ships flat and unfolds to reveal openings used to hold tubes upright inside storage boxes (sold separately). Divders will fit into standard 2 inch and 3 inch high storage boxes.
Catalog No. 50-998-924 Supplier Research Products International Corp Supplier No. 181034
Cardboard cell divider is designed for use with cardboard storage boxes and cardboard freezer boxes.
Cell divider ships flat and unfolds to reveal openings used to hold tubes upright inside storage boxes (sold separately). Divders will fit into standard 2 inch and 3 inch high storage boxes.