Learn More
Alkali Scientific Alkali Scientific™ Cell Divider for 4" PCR Tube Freezer Box, 64, 1/pk
SDP
Catalog No. 503026546
Encompass_Preferred
Cardboard Material Perfect for Microtubes Additional box support and crush resistance Reusable and recyclable Makes packing and unpacking easy These economical cryo/freezer storage Cell Box Divider offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation. Choose from a wide range of boxes and dividers to accommodate tubes from 0.5 to 50 mL
Catalog No. 50-302-6546 Supplier Alkali Scientific Supplier No. CD464
Cardboard Material Perfect for Microtubes Additional box support and crush resistance Reusable and recyclable Makes packing and unpacking easy These economical cryo/freezer storage Cell Box Divider offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation. Choose from a wide range of boxes and dividers to accommodate tubes from 0.5 to 50 mL