Learn More
Research Products International Corp Cell Dividers for Cardboard Storage Box, Capacity 16, 50 ml Tubes
SDP
Catalog No. 50550275
Encompass
Cardboard cell dividers for use with RPI Storage Boxes. Fits boxes sized 5 3/4 x 5 3/4 inches and holds up to 16 x 50 ml tubes. Dividers come collapsed for shipping and would need to be expanded before use in Storage Boxes.
Catalog No. 50-550-275 Supplier Research Products International Corp Supplier No. 247572
Cardboard cell dividers for use with RPI Storage Boxes. Fits boxes sized 5 3/4 x 5 3/4 inches and holds up to 16 x 50 ml tubes. Dividers come collapsed for shipping and would need to be expanded before use in Storage Boxes.