Learn More
Research Products International Corp Cell Dividers for Cardboard Storage Box, Capacity 36, 15ml Tubes
SDP
Catalog No. 50550276
Encompass
Cardboard cell dividers for use with RPI Storage Boxes. Fits boxes sized 5 3/4 x 5 3/4 inches and holds up to 36 x 15 ml tubes. Dividers come collapsed for shipping and would need to be expanded before use in Storage Boxes.
Catalog No. 50-550-276 Supplier Research Products International Corp Supplier No. 247573
Cardboard cell dividers for use with RPI Storage Boxes. Fits boxes sized 5 3/4 x 5 3/4 inches and holds up to 36 x 15 ml tubes. Dividers come collapsed for shipping and would need to be expanded before use in Storage Boxes.