Learn More
Andwin Scientific Cell Dividers 9 x 9, 12 pack
SDP
Catalog No. NC9415172
Encompass_Preferred
The TrueNorth cardboard cryovial box cell divider provides secure and organized storage for small samples and 1.5-2 mL tubes. There are 81 wells that have a diameter of 11.5 mm. This product is specially designed with vent holes at the bottom to permit fast circulation and disposal of cold air, vapor, or LN. This product is sold in a pack of 12.
Catalog No. NC9415172 Supplier Andwin Scientific Supplier No. TX2860H
The TrueNorth cardboard cryovial box cell divider provides secure and organized storage for small samples and 1.5-2 mL tubes. There are 81 wells that have a diameter of 11.5 mm. This product is specially designed with vent holes at the bottom to permit fast circulation and disposal of cold air, vapor, or LN. This product is sold in a pack of 12.